siRNA Details
Gene Symbol: Pou5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009178.2 | NP_001009178.1 | 5 | 926 | ||
GenBank mRNA | BC158566.1 | - | 5 | 926 | 9092936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
