siRNA Details
Gene Symbol: S100z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191701.1 | NP_001178630.1 | 2 | 260 | ||
XM_008760662.1 | XP_008758884.1 | 4 | 1052 | |||
XM_008760663.1 | XP_008758885.1 | 4 | 1153 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
