siRNA Details
Gene Symbol: Card6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106413.1 | NP_001099883.1 | 3 | 2468 | ||
XM_008760760.1 | XP_008758982.1 | 3 | 2595 | |||
XM_008760761.1 | XP_008758983.1 | 4 | 2518 |
