siRNA Details
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106414.1 | NP_001099884.1 | 4, 5 | 639 | ||
XM_006231993.2 | XP_006232055.1 | 4, 5 | 528 | |||
XM_006231994.1 | XP_006232056.1 | 4, 5 | 509 |

More Information
Other Species Targeted

