siRNA Details
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106418.1 | NP_001099888.1 | 3 | 329 | ||
XM_006257698.2 | XP_006257760.1 | 3 | 213 | |||
XM_006257699.2 | XP_006257761.1 | 3 | 187 | |||
XM_008773706.1 | XP_008771928.1 | 2 | 2125 |
