siRNA Details
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760826.1 | XP_008759048.1 | 12 | 1285 | ||
XM_215517.8 | XP_215517.5 | 12 | 1230 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760826.1 | XP_008759048.1 | 12 | 1285 | ||
XM_215517.8 | XP_215517.5 | 12 | 1230 |