siRNA Details
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760826.1 | XP_008759048.1 | 4 | 552 | ||
XM_215517.8 | XP_215517.5 | 4 | 497 |

More Information
Other Species Targeted

Gene Ontology Categories:
