siRNA Details
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004078.1 | NP_001004078.1 | 3 | 246 | ||
GenBank mRNA | AY683456.1 | - | 2 | 128 | ||
BC059165.1 | - | 3 | 316 | |||
BC079441.1 | - | 3 | 246 | 7115414 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: Z31555.
Gene Ontology Categories:
