siRNA Details
Gene Symbol: Ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024978.1 | NP_001020149.1 | 5 | 437 | ||
XM_006232123.2 | XP_006232185.1 | 6 | 762 | |||
XM_006232126.2 | XP_006232188.1 | 6 | 1259 | |||
XM_008760852.1 | XP_008759074.1 | 6 | 762 | |||
XM_008760853.1 | XP_008759075.1 | 6 | 1259 | |||
GenBank mRNA | BC092614.1 | - | 5 | 437 | 7311942 |

More Information
Other Species Targeted

Gene Ontology Categories:
