siRNA Details
Gene Symbol: tGap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007635.2 | NP_001007636.2 | 23, 24 | 3418 | ||
XM_006232134.2 | XP_006232196.1 | 25, 26 | 3760 | |||
GenBank mRNA | AY631396.1 | - | 23, 24 | 3418 | ||
AY631399.1 | - | 23, 24 | 3183 | |||
AY631400.1 | - | 19, 20 | 2557 |
