siRNA Details
Gene Symbol: Tpd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106421.1 | NP_001099891.1 | 3, 4 | 433 | ||
XM_006232140.1 | XP_006232202.1 | 3, 4 | 697 | |||
XM_006232141.1 | XP_006232203.1 | 3, 4 | 697 | |||
XM_006232142.1 | XP_006232204.1 | 3, 4 | 697 | |||
XM_006232143.2 | XP_006232205.1 | 3, 4 | 1086 | |||
XM_008760861.1 | XP_008759083.1 | 3, 4 | 697 | |||
XM_008760862.1 | XP_008759084.1 | 3, 4 | 1086 | |||
XM_008760863.1 | XP_008759085.1 | 3, 4 | 1086 | |||
GenBank mRNA | GQ499323.1 | - | 3, 4 | 400 |
