siRNA Details
Gene Symbol: Tnik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106422.1 | NP_001099892.1 | 25 | 3480 | ||
XM_008760893.1 | XP_008759115.1 | 24 | 4125 | |||
XM_008760894.1 | XP_008759116.1 | 24 | 4125 | |||
XM_008760895.1 | XP_008759117.1 | 23 | 4101 | |||
XM_008760896.1 | XP_008759118.1 | 23 | 4038 | |||
XM_008760897.1 | XP_008759119.1 | 22 | 4013 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
