siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106423.1 | NP_001099893.1 | 13 | 3575 | ||
XM_006232249.2 | XP_006232311.1 | 13 | 3383 | |||
XM_006232250.2 | XP_006232312.1 | 13 | 3625 | |||
XM_008760907.1 | XP_008759129.1 | 13 | 3380 | |||
XM_008760908.1 | XP_008759130.1 | 12 | 3356 | |||
XM_008760909.1 | XP_008759131.1 | 12 | 3352 | |||
XM_008760910.1 | XP_008759132.1 | 13 | 3382 | |||
XM_008760911.1 | XP_008759133.1 | 12 | 2380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
