siRNA Details
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106423.1 | NP_001099893.1 | 7 | 2772 | ||
XM_006232249.2 | XP_006232311.1 | 7 | 2580 | |||
XM_006232250.2 | XP_006232312.1 | 7 | 2822 | |||
XM_008760907.1 | XP_008759129.1 | 7 | 2577 | |||
XM_008760908.1 | XP_008759130.1 | 7 | 2580 | |||
XM_008760909.1 | XP_008759131.1 | 7 | 2576 | |||
XM_008760910.1 | XP_008759132.1 | 7 | 2579 | |||
XM_008760911.1 | XP_008759133.1 | 7 | 1604 |

More Information
Other Species Targeted

Gene Ontology Categories:
