siRNA Details
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013910.2 | NP_001013932.1 | 3 | 327 | ||
GenBank mRNA | AY552804.1 | - | 3 | 254 | ||
BC097370.1 | - | 3 | 327 | 7444022 | ||
DQ120489.1 | - | 3 | 219 | |||
DQ120490.1 | - | 3 | 219 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: M63658.
Gene Ontology Categories:
