siRNA Details
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181768.2 | NP_861433.2 | 7 | 756 | ||
XM_006232269.1 | XP_006232331.1 | 7 | 836 | |||
XM_006232270.2 | XP_006232332.1 | 7 | 836 | |||
GenBank mRNA | BC162016.1 | - | 7 | 756 | 9092768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
