siRNA Details
Gene Symbol: Adad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006977.2 | NP_001006978.2 | 11 | 1680 | ||
XM_006232278.2 | XP_006232340.1 | 11 | 1864 | |||
XM_006232279.2 | XP_006232341.1 | 12 | 1716 | |||
GenBank mRNA | BC105906.1 | - | 11 | 1746 | 7453283 |

More Information
Other Species Targeted

Gene Ontology Categories:
