siRNA Details
Gene Symbol: Spry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC169059.1 | - | 3 | 1376 | 7938818 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011896, BC053428, AK140405, AF176903, BC039139, and others.
