siRNA Details
Gene Symbol: LOC499607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760983.1 | XP_008759205.1 | 17 | 2782 |
Gene Symbol: LOC689629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760982.1 | XP_008759204.1 | 32 | 5057 |
