siRNA Details
Gene Symbol: Pcdh18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100524.1 | NP_001093994.1 | 1 | 1134 | ||
XM_006232322.2 | XP_006232384.1 | 1 | 1199 | |||
GenBank mRNA | BC168716.1 | - | 1 | 1134 | 9099515 |
