siRNA Details
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106444.1 | NP_001099914.1 | 5 | 816 | ||
GenBank mRNA | BC167108.1 | - | 5 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
