siRNA Details
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106444.1 | NP_001099914.1 | 3 | 716 | ||
GenBank mRNA | BC167108.1 | - | 3 | 277 |

More Information
Other Species Targeted

Gene Ontology Categories:
