siRNA Details
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106449.2 | NP_001099919.1 | 8 | 941 | ||
NM_001270650.1 | NP_001257579.1 | 9 | 1015 | |||
NM_001270651.1 | NP_001257580.1 | 9 | 988 | |||
NM_001270652.1 | NP_001257581.1 | 8 | 918 | |||
NM_001270653.1 | NP_001257582.1 | 8 | 945 | |||
XM_006232855.2 | XP_006232917.1 | 9 | 1010 | |||
XM_008761282.1 | XP_008759504.1 | 8 | 976 | |||
GenBank mRNA | FM111895.1 | - | 5 | 397 | ||
FQ214903.1 | - | 8 | 878 | |||
FQ215988.1 | - | 9 | 944 | |||
FQ216125.1 | - | 8 | 867 | |||
FQ216236.1 | - | 6 | 612 | |||
FQ216249.1 | - | 9 | 938 | |||
FQ217203.1 | - | 8 | 865 |
