siRNA Details
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106449.2 | NP_001099919.1 | 2 | 217 | ||
NM_001270650.1 | NP_001257579.1 | 3 | 291 | |||
NM_001270651.1 | NP_001257580.1 | 3 | 264 | |||
NM_001270652.1 | NP_001257581.1 | 2 | 194 | |||
NM_001270653.1 | NP_001257582.1 | 2 | 221 | |||
XM_006232855.2 | XP_006232917.1 | 3 | 286 | |||
XM_008761282.1 | XP_008759504.1 | 2 | 252 | |||
GenBank mRNA | FN800735.1 | - | 2 | 175 | ||
FQ214903.1 | - | 2 | 154 | |||
FQ215988.1 | - | 3 | 220 | |||
FQ216125.1 | - | 2 | 143 | |||
FQ216249.1 | - | 3 | 214 | |||
FQ216534.1 | - | 2 | 120 | |||
FQ217203.1 | - | 2 | 141 | |||
FQ224853.1 | - | 2 | 142 |
