siRNA Details
Gene Symbol: Ankrd34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024980.1 | NP_001020151.1 | 2 | 334 | ||
XM_006232953.2 | XP_006233015.1 | 3 | 1696 | |||
XM_008761283.1 | XP_008759505.1 | 2 | 1942 | |||
XM_008761284.1 | XP_008759506.1 | 3 | 1841 | |||
GenBank mRNA | BC091343.1 | - | 2 | 334 | 7315500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
