siRNA Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271138.1 | NP_001258067.1 | 5 | 416 | ||
XM_006232956.2 | XP_006233018.1 | 5 | 414 | |||
GenBank mRNA | DQ359102.1 | - | 5 | 368 | ||
FQ210627.1 | - | 5 | 389 |
