siRNA Details
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134959.1 | NP_001128431.1 | 19 | 2360 | ||
XM_008761138.1 | XP_008759360.1 | 19 | 2568 | |||
XM_008761139.1 | XP_008759361.1 | 17 | 2398 | |||
XM_008761140.1 | XP_008759362.1 | 19 | 2567 | |||
GenBank mRNA | BC168665.1 | - | 19 | 2360 | 7933243 | |
FQ226108.1 | - | 19 | 2406 | |||
FQ231267.1 | - | 19 | 2406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
