siRNA Details
Gene Symbol: Etv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106450.1 | NP_001099920.1 | 4 | 831 | ||
XM_006232620.2 | XP_006232682.1 | 5 | 922 | |||
XM_006232621.2 | XP_006232683.1 | 5 | 1539 | |||
XM_006232622.1 | XP_006232684.1 | 4 | 1089 | |||
XM_008761141.1 | XP_008759363.1 | 5 | 1241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
