siRNA Details
Gene Symbol: Rhoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106461.1 | NP_001099931.1 | 2 | 383 | ||
XM_006233074.1 | XP_006233136.1 | 2 | 357 | |||
GenBank mRNA | BC166849.1 | - | 2 | 399 | 9088252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
