siRNA Details
Gene Symbol: Eps8l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106463.1 | NP_001099933.1 | 9 | 980 |
Gene Symbol: LOC100912384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773861.1 | XP_008772083.1 | 9 | 1134 |
