siRNA Details
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017449.1 | NP_001017449.1 | 3 | 564 | ||
XM_006233214.1 | XP_006233276.1 | 3 | 377 | |||
XM_008761454.1 | XP_008759676.1 | 2 | 425 | |||
XM_008761455.1 | XP_008759677.1 | 2 | 426 | |||
GenBank mRNA | BC092598.1 | - | 3 | 564 | 7321341 |
