siRNA Details
Gene Symbol: Myoz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233264.2 | XP_006233326.1 | 3 | 412 | ||
GenBank mRNA | FQ215232.1 | - | 3 | 307 | ||
FQ215285.1 | - | 3 | 337 | |||
FQ215331.1 | - | 3 | 366 | |||
FQ215936.1 | - | 3 | 307 | |||
FQ216537.1 | - | 3 | 368 | |||
FQ216608.1 | - | 3 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
