siRNA Details
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191716.1 | NP_001178645.1 | 9 | 2228 | ||
XM_006233268.2 | XP_006233330.1 | 10 | 2899 |

More Information
Other Species Targeted

Gene Ontology Categories:
