siRNA Details
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191716.1 | NP_001178645.1 | 6 | 1718 | ||
XM_006233268.2 | XP_006233330.1 | 7 | 2389 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191716.1 | NP_001178645.1 | 6 | 1718 | ||
XM_006233268.2 | XP_006233330.1 | 7 | 2389 |