siRNA Details
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106473.1 | NP_001099943.1 | 4 | 396 | ||
XM_006233322.2 | XP_006233384.1 | 4 | 305 | |||
GenBank mRNA | BC095848.1 | - | 3 | 790 |

More Information
Other Species Targeted

Gene Ontology Categories:
