siRNA Details
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106474.1 | NP_001099944.1 | 8 | 1766 | ||
XM_006233275.2 | XP_006233337.1 | 8 | 1842 | |||
XM_006233276.2 | XP_006233338.1 | 7 | 1736 | |||
XM_006233277.2 | XP_006233339.1 | 6 | 1562 | |||
XM_008761458.1 | XP_008759680.1 | 8 | 1839 |

More Information
Other Species Targeted

Gene Ontology Categories:
