siRNA Details
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106474.1 | NP_001099944.1 | 12 | 2300 | ||
XM_006233275.2 | XP_006233337.1 | 12 | 2376 | |||
XM_006233276.2 | XP_006233338.1 | 11 | 2270 | |||
XM_006233277.2 | XP_006233339.1 | 10 | 2096 | |||
XM_008761458.1 | XP_008759680.1 | 12 | 2373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
