siRNA Details
Gene Symbol: Clcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106479.1 | NP_001099949.1 | 21 | 2386 | ||
XM_006239377.2 | XP_006239439.1 | 20 | 4334 | |||
XM_006239378.2 | XP_006239440.1 | 21 | 2718 |
