siRNA Details
Gene Symbol: Clcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106479.1 | NP_001099949.1 | 3, 4 | 236 | ||
XM_006239377.2 | XP_006239439.1 | 2, 3 | 2184 | |||
XM_006239378.2 | XP_006239440.1 | 3, 4 | 568 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011929, AK165362, AK145053.
Gene Ontology Categories:
