siRNA Details
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178092.2 | NP_835193.1 | 10 | 1226 | ||
XM_006234275.2 | XP_006234337.1 | 10 | 1777 | |||
XM_006234276.2 | XP_006234338.1 | 10 | 1774 | |||
XM_006234277.2 | XP_006234339.1 | 10 | 1557 | |||
XM_006234279.2 | XP_006234341.1 | 10 | 1772 | |||
XM_006234280.2 | XP_006234342.1 | 9 | 1684 | |||
XM_006234282.2 | XP_006234344.1 | 9 | 1467 | |||
XM_006234283.2 | XP_006234345.1 | 9 | 1464 | |||
XM_006234284.2 | XP_006234346.1 | 9 | 1683 | |||
XM_006234285.2 | XP_006234347.1 | 9 | 1466 | |||
GenBank mRNA | AF439855.1 | - | 9 | 1136 | ||
AF439856.1 | - | 10 | 1226 | |||
AY579373.1 | - | 10 | 1229 | |||
AY579374.1 | - | 9 | 1139 | |||
AY579375.1 | - | 9 | 1139 | |||
AY579376.1 | - | 9 | 1139 | |||
AY579377.1 | - | 9 | 1139 | |||
JX073715.1 | - | 9 | 1183 | |||
JX073716.2 | - | 10 | 1270 | |||
JX073717.1 | - | 9 | 1183 | |||
JX073718.2 | - | 10 | 1270 | |||
KF305251.1 | - | 10 | 1195 | |||
KF736947.1 | - | 10 | 1161 | |||
KF736948.1 | - | 9 | 1071 | |||
KF736949.1 | - | 10 | 1194 | |||
KF736950.1 | - | 9 | 1104 | |||
KJ452197.1 | - | 11 | 1210 |
