siRNA Details
Gene Symbol: Ube2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047857.3 | NP_001041322.2 | 2 | 531 | ||
XM_006234405.2 | XP_006234467.1 | 2 | 458 | |||
GenBank mRNA | DQ480743.1 | - | 2 | 527 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009454, AK168072, BC011477, X92664, AK076011, and others.
Gene Ontology Categories:
