siRNA Details
Gene Symbol: Prg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106487.1 | NP_001099957.1 | 4 | 600 | ||
XM_006234449.1 | XP_006234511.1 | 5 | 636 | |||
XM_006234450.1 | XP_006234512.1 | 5 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
