siRNA Details
Gene Symbol: Olr526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001375.1 | NP_001001375.1 | 1 | 625 |
Gene Symbol: Olr530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001376.1 | NP_001001376.1 | 1 | 625 |
