siRNA Details
Gene Symbol: Olr669 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000349.1 | NP_001000349.1 | 1 | 795 |
Gene Symbol: Olr666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000924.1 | NP_001000924.1 | 1 | 795 |
