siRNA Details
Gene Symbol: Olr679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000354.1 | NP_001000354.1 | 1 | 339 |
Gene Symbol: Olr681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000631.1 | NP_001000631.1 | 1 | 339 |

More Information
Other Species Targeted

Gene Ontology Categories:
