siRNA Details
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106490.1 | NP_001099960.1 | 29 | 3506 | ||
XM_006234503.2 | XP_006234565.1 | 31 | 3562 | |||
XM_006234504.1 | XP_006234566.1 | 31 | 3562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
