siRNA Details
Gene Symbol: Chst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011955.1 | NP_001011955.1 | 3 | 683 | ||
XM_006234549.2 | XP_006234611.1 | 2 | 1524 | |||
XM_008762023.1 | XP_008760245.1 | 3 | 632 | |||
GenBank mRNA | BC086551.1 | - | 3 | 683 | 7302418 | |
FQ211655.1 | - | 3 | 1012 |

More Information
Other Species Targeted

Gene Ontology Categories:
