siRNA Details
Gene Symbol: RGD1309730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106491.2 | NP_001099961.2 | 3 | 302 | ||
GenBank mRNA | BC166899.1 | - | 3 | 302 | 6922132 | |
FQ214875.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
