siRNA Details
Gene Symbol: RGD1309730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106491.2 | NP_001099961.2 | 8 | 893 | ||
GenBank mRNA | BC166899.1 | - | 8 | 893 | 6922132 | |
FQ214875.1 | - | 8 | 923 |

More Information
Other Species Targeted

