siRNA Details
Gene Symbol: Ehf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106493.2 | NP_001099963.1 | 6, 7 | 681 | ||
XM_006234639.2 | XP_006234701.1 | 6, 7 | 3180 | |||
GenBank mRNA | BC166742.1 | - | 6, 7 | 681 | 7937842 |

More Information
Other Species Targeted

Gene Ontology Categories:
